Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCN4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1691R

Description
HCN4 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. It has been shown that HCN4 channels function as receptors for sour taste, and are associated with pacemaker potential generation in the sinoatrial node.Specifications
HCN4 | |
Polyclonal | |
Unconjugated | |
Hcn4 | |
Bcng3; BCNG-3; brain cyclic nucleotide-gated channel 3; Hcn3; HCN4; hyperpolarization activated cyclic nucleotide gated potassium channel 4; hyperpolarization activated cyclic nucleotide-gated cation channel 4; hyperpolarization activated cyclic nucleotide-gated potassium channel 4; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4; SSS2 | |
Rabbit | |
Protein A | |
RUO | |
10021 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9Y3Q4 | |
Hcn4 | |
KLH conjugated synthetic peptide derived from human HCN4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction