Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCN4 Rat anti-Mouse, Unconjugated, Clone: PG2-1A4, MilliporeSigma™

Catalog No. MABN187125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1871-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1871-25 Supplier MilliporeSigma Supplier No. MABN187125UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-HCN4, clone PG2-1A4 Antibody, Cat. No. MABN1871, is a highly specific rat monoclonal antibody that targets murine HCN4 and has been tested for use in immunofluorescence, Immunohistochemistry, Electron Microscopy, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCN4
Applications Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Western Blot
Classification Monoclonal
Clone PG2-1A4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4;Brain cyclic nucleotide-gated channel 3;BCNG-3
Gene Symbols Hcn4;Bcng3
Host Species Rat
Immunogen GST-tagged recombinant fragment corresponding to 70 amino acids from the C-terminal region of murine Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4 (HCN4).
Molecular Weight of Antigen 127.41 kDa calculated.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone PG2-1A4 detects murine Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4 (HCN4). It targets an epitope within 70 amino acids from the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.