Learn More
HCoV-229E Spike RBD Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Spike RBD |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2840H |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P15423.1 |
| Host Species | Rabbit |
| Immunogen | Human embryonic kidney cell, HEK293-derived HCoV-229E Spike RBD, Ser292-Asp453, Accession # P15423.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.