Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCoV-HKU1 Human Coronavirus Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $420.50
Specifications
| Antigen | Spike RBD |
|---|---|
| Clone | 1045307 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10936SP
![]() |
R&D Systems
MAB10936SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10936100
![]() |
R&D Systems
MAB10936100 |
100 μg |
Each for $420.50
|
|
|||||
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in HCoV-HKU1 samples. It is validated for ImmunocytochemistrySpecifications
| Spike RBD | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Human embryonic kidney cell HEK293-derived HCoV-HKU1 Spike RBD, Thr310-Tyr624, Accession # Q5MQD0.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1045307 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| HCoV-HKU1 | |
| Q5MQD0.1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title