Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCoV-HKU1 Nucleoprotein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120767 Supplier Invitrogen™ Supplier No. PA5120767
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Human coronavirus (HCoV-HKU1) Nucleocapsid Protein (His Tag). Immunogen sequence: WYRHSRRSFK TADGQQKQLL PRWYFYYLGT GPYANASYGE SLEGVFWVAN HQADTSTPSD VSSRDPTTQE AIPTRFPPGT ILPQGYYVEG SGRSASNSRP G.

Packages the positive strand viral genome RNA into a helical ribonucleocapsid (RNP) and plays a fundamental role during virion assembly through its interactions with the viral genome and membrane protein M. Plays an important role in enhancing the efficiency of subgenomic viral RNA transcription as well as viral replication. [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCoV-HKU1 Nucleoprotein
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.642 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Alias Coronavirus; HCoV; HKU1; Nucleoprotein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of coronavirus Nucleoprotein (YP_1732421).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.