Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCoV-HKU1 Spike S1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120768 Supplier Invitrogen™ Supplier No. PA5120768
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Human coronavirus HKU1 (isolate N5) (HCoV-HKU1) Spike Protein (S1+S2 ECDHis Tag). Immunogen sequence: NGIFSRVKNT KLYVNKTLYS EFSTIVIGSV FINNSYTIVV QPHNGVLEIT ACQYTMCEYP HTICKSKGSS RNESWHFDKS EPLCLFKKNF TYNVSTDWLY F.

The S1 spike protein attaches the virion to the cell membrane by interacting with host receptor, initiating the infection. [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCoV-HKU1 Spike S1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Alias Coronavirus; HCoV; HKU1; S1; Spike
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.