Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCoV-OC43 Nucleocapsid Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
$177.10 - $454.00
Specifications
Antigen | Nucleocapsid |
---|---|
Clone | 1051436 |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11289100
![]() |
R&D Systems
MAB11289100 |
100 μg |
Each for $454.00
|
|
|||||
MAB11289SP
![]() |
R&D Systems
MAB11289SP |
25 μg |
Each for $177.10
|
|
|||||
Description
Nucleocapsid Monoclonal antibody specifically detects Nucleocapsid in HCoV-OC43 samples. It is validated for Western Blot, Flow Cytometry, ImmunocytochemistrySpecifications
Nucleocapsid | |
Western Blot 1 ug/mL, Flow Cytometry 0.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Sf-21 (baculovirus)-derived hcov-oc43 Nucleocapsid, Met1-Ile448, Accession # YP_009555245.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1051436 | |
Western Blot, Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
HCoV-OC43 | |
YP_009555245.1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title