Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCoV-OC43 Spike S1 Recombinant Rabbit Monoclonal Antibody (5M10C7)
GREENER_CHOICE

Catalog No. FEMA551285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551285 Supplier Invitrogen™ Supplier No. MA551285
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

HCoV-OC43 Spike S1 Recombinant Monoclonal Antibody for ELISA, Western Blot

The S1 spike protein attaches the virion to the cell membrane by interacting with host receptor, initiating the infection. [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCoV-OC43 Spike S1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5M10C7
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Alias Coronavirus; HCoV; OC43; S1; Spike
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.