Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HCoV-OC43 Spike S1 Recombinant Rabbit Monoclonal Antibody (8Y5I1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551461
Description
HCoV-OC43 Spike S1 Recombinant Monoclonal Antibody for Western Blot, ELISA
The S1 spike protein attaches the virion to the cell membrane by interacting with host receptor, initiating the infection. [Uniprot].
Specifications
HCoV-OC43 Spike S1 | |
Recombinant Monoclonal | |
0.41 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Virus | |
Antibody | |
Liquid |
ELISA, Western Blot | |
8Y5I1 | |
Unconjugated | |
Coronavirus; HCoV; OC43; S1; Spike | |
A synthesized peptide derived from human HCoV-OC43 Spike S1. | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction