Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983757
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3757 Supplier Bioss Supplier No. BS15428R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Has been shown to act both as an intestinal proton-coupled high-affinity folate transporter and as an intestinal heme transporter which mediates heme uptake from the gut lumen into duodenal epithelial cells. The iron is then released from heme and may be transported into the bloodstream. Dietary heme iron is an important nutritional source of iron. Shows a higher affinity for folate than heme.

Specifications

Antigen HCP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC46A1
Gene Accession No. Q96NT5
Gene Alias 1110002C08Rik; BOS_18549; D11Ertd18e; G21; Hcp1; Heme carrier protein 1; LOC100009317; Pcft; PCFT/HCP1; Proton-coupled folate transporter; retinal pigment epithelilum transporter; Retinal pigment epithelium transporter; RGD1309472; Slc46a1; solute carrier family 46 (folate transporter), member 1; Solute carrier family 46 member 1; solute carrier family 46, member 1; TRPE
Gene Symbols SLC46A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HCP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113235
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.