Learn More
StatLab HCS Control Slides AFB Adhesion Slides
SDP
Catalog No. 23640163
Click to view available options
Quantity:
100/Pk.
25/Pk.
HCS Control Slides AFB Adhesion Slides
  • HCS Control Slides consist of formalin fixed, paraffin embedded tissue sections which contain known positive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • StatLab HCS control slides are created using adhesion white glass slides with 90° corners
  • Each package includes a specific staining protocol
Specifications
Type Control Slide
Quantity 25/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.