Learn More
HCS Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304261A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Holocarboxylase synthetase (EC 6.3.4.10) covalently links biotin to propionyl-CoA carboxylase (PCCA; MIM 232000), pyruvate carboxylase (PC; MIM 608786), alpha-methylcrotonyl-CoA carboxylase (MCCC1; MIM 609010), and acetyl-CoA carboxylase (ACACA; MIM 200350).Specifications
HCS | |
Polyclonal | |
Unconjugated | |
HLCS | |
HCS, biotin apo-protein ligase, biotin--[acetyl-CoA-carboxylase] ligase, biotin--[methylcrotonoyl-CoA-carboxylase] ligase, biotin--[methylmalonyl-CoA-carboxytransferase] ligase, biotin--protein ligase, holocarboxylase synthetase (biotin-(proprionyl-Coenzyme A-carbo | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3141 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P50747 | |
HLCS | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.