Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hCV-2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543844
Description
Peptide sequence: NGHKRDDLIG GDGNFKFDVD DFAESWRVES NEFCNRPQRK PVPELCQGTV Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 85%; Zebrafish: 79%.
BMPER contains 1 TIL (trypsin inhibitory-like) domain, 5 VWFC domains and 1 VWFD domain.BMPER is the inhibitor of bone morphogenetic protein (BMP) functions. It may regulate BMP responsiveness of osteoblasts and chondrocytes.
Specifications
hCV-2 | |
Polyclonal | |
Unconjugated | |
Bmper | |
3110056H04Rik; BMP binding endothelial regulator; BMP-binding endothelial regulator; BMP-binding endothelial regulator precursor protein; BMP-binding endothelial regulator protein; Bmper; Bone morphogenetic protein-binding endothelial cell precursor-derived regulator; CRIM3; crossveinless 2; crossveinless-2; Cv2; CV-2; cvl2; hCV2; id:ibd5071; KIAA1965; mCV2; Protein crossveinless-2; RGD1563373; secreted modulator of bone morphogenetic proteins | |
Rabbit | |
Affinity chromatography | |
RUO | |
168667 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N8U9 | |
Bmper | |
Synthetic peptide directed towards the C-terminal of human BMPER (aa 501-550). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction