Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV Core Protein Mouse anti-Virus, Alexa Fluor™ 488, Clone: C7-50, BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569372
Description
HCV Core Protein Monoclonal antibody specifically detects HCV Core Protein in Virus samples. It is validated for Intracellular staining (flow cytometry)
Specifications
HCV Core Protein | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
951475 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Flow Cytometry, Control | |
C7-50 | |
Alexa Fluor 488 | |
HCV Core Protein | |
Purified HCV Core Recombinant Protein | |
0.1 mg | |
Primary | |
Virus | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction