Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCV E1E2 Human anti-Virus, Clone: AR5A, MilliporeSigma™

Catalog No. MABF2334100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2334100 100 μL
MABF233425 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2334100 Supplier MilliporeSigma Supplier No. MABF2334100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

HCV E1E2 Monoclonal antibody specifically detects HCV E1E2 in Virus samples. It is validated for ELISA, Immunoprecipitation, Neutralizing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCV E1E2
Applications ELISA, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone AR5A
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified human recombinant monoclonal antibody in PBS without preservatives.
Host Species Human
Immunogen Antibody Fab fragment V1 expressed in E. coli, purified, and converted into full-length IgG1 molecules in CHO-K1 cells.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.