Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV E2 Human anti-Virus, Clone: AR1B, MilliporeSigma™
Human Monoclonal Antibody
$182.31 - $463.74
Specifications
Antigen | HCV E2 |
---|---|
Clone | AR1B |
Concentration | 1.0 mg/mL |
Applications | ELISA, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF233525
|
MilliporeSigma
MABF233525UG |
25 μg |
Each for $182.31
|
|
|||||
MABF2335100
|
MilliporeSigma
MABF2335100UG |
100 μg |
Each for $463.74
|
|
|||||
Description
HCV E2 Monoclonal antibody specifically detects HCV E2 in Virus samples. It is validated for ELISA, Neutralizing.Specifications
HCV E2 | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified human recombinant monoclonal antibody in PBS without azide. | |
IgG1 | |
Protein G purified |
AR1B | |
ELISA, Neutralization | |
Unconjugated | |
Human | |
Virus | |
Antibody Fab fragment D1 expressed in E. coli, purified, and converted into full-length IgG1 molecules in CHO-K1 cells. | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title