Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCV E2 Mouse anti-Hepatitis C virus, Unconjugated, Clone: 19B3, MilliporeSigma™

Catalog No. MABF23081MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF23081MI 100 μg
MABF23082MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF23081MI Supplier MilliporeSigma Supplier No. MABF2308100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HCV E2 Monoclonal antibody specifically detects HCV E2 in Hepatitis C virus samples. It is validated for ELISA, Neutralizing, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCV E2
Applications ELISA, Neutralization, Western Blot
Classification Monoclonal
Clone 19B3
Conjugate Unconjugated
Gene Alias Hepatittis C virus E2 protein
Host Species Mouse
Immunogen DNA constructs of E1E2 followed by a boost with truncated E2 protein (aa 412-645).
Molecular Weight of Antigen 327.15 kDa calculated for the entire genome polyprotein.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 19B3 is a mouse monoclonal antibody that detects the envelope glycoprotein E2 of the Hepatitis C virus. It targets an epitope within 12 amino acids from the N-terminal region of the E2.
Target Species Hepatitis C virus
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.