Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV-NS3 Antibody (1B6), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280122UV
This item is not returnable.
View return policy
Description
HCV-NS3 Monoclonal antibody specifically detects HCV-NS3 in Virus samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
HCV-NS3 | |
Monoclonal | |
DyLight 350 | |
Nonstructural Protein 3, NS3 protease, Q91RS4 | |
Recombinant HCV-NS3 serine protease domain. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Immunofluorescence | |
1B6 | |
50mM Sodium Borate | |
Mouse | |
>95%, by SDS-PAGE | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction