Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV-NS3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313013
This item is not returnable.
View return policy
Description
HCV-NS3 Polyclonal antibody specifically detects HCV-NS3 in Virus samples. It is validated for Western BlotSpecifications
| HCV-NS3 | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Nonstructural Protein 3, NS3 protease, Q91RS4 | |
| Recombinant protein encompassing a sequence within the C-terminus region of HCV-NS3. (HCV 2a (strain JFH-1)). The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction