Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV NS5A transactivated protein 8 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8158R

Description
The C-terminus of BTBD1 binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
HCV NS5A transactivated protein 8 | |
Polyclonal | |
Unconjugated | |
BTBD1 | |
1190005H08Rik; BOS_20177; BTB (POZ) domain containing 1; BTB domain containing 1; BTB/POZ domain-containing protein 1; Btbd1; C15orf1; Glucose signal-repressing protein; Gsrp; HCV NS5A-transactivated protein 8; Hepatitis C virus NS5A-transactivated protein 8; NS5ATP8 | |
Rabbit | |
Protein A | |
RUO | |
53339 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9H0C5 | |
BTBD1 | |
KLH conjugated synthetic peptide derived from human BTBD1/HCV NS5A transactivated protein 8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction