Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV-NS5b Antibody (5B-12B7), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280124G
This item is not returnable.
View return policy
Description
HCV-NS5b Monoclonal antibody specifically detects HCV-NS5b in Virus samples. It is validated for ImmunofluorescenceSpecifications
HCV-NS5b | |
Monoclonal | |
DyLight 488 | |
EC:2.7.7.48, Nonstructural Protein 5B, NS5B, Q8JXY1, RNA-directed RNA polymerase | |
Recombinant HCV-NS5b RNA-dependent RNA polymerase. | |
0.1 mL | |
Virology Bacteria and Parasites | |
951475 | |
Store at 4°C in the dark. | |
IgG2a |
Immunofluorescence | |
5B-12B7 | |
50mM Sodium Borate | |
Mouse | |
>95%, by SDS-PAGE | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction