Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV-NS5b Antibody (5B-3B1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280123
Description
HCV-NS5b Monoclonal specifically detects HCV-NS5b in Virus samples. It is validated for Western Blot.Specifications
HCV-NS5b | |
Monoclonal | |
1 mg/mL | |
Western Blot | |
EC:2.7.7.48, Nonstructural Protein 5B, NS5B, Q8JXY1, RNA-directed RNA polymerase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes HCV-NS5b (epitope mapped to NS5B aa 372-382). It recognizes genotype 1 a and also 1b (albeit with lower affinity). It does not recognize genotype 2a. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot | |
5B-3B1 | |
Unconjugated | |
PBS containing 10% glycerol | |
POLY | |
Recombinant HCV-NS5b RNA-dependent RNA polymerase (genotype 1a). | |
100 μg | |
Virology Bacteria and Parasites | |
951475 | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction