Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HD-polyCys-Ct Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN167110MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN167110MI 100 μL
ABN167125MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN167110MI Supplier MilliporeSigma Supplier No. ABN1671100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HD-polyCys-Ct Polyclonal antibody specifically detects HD-polyCys-Ct in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HD-polyCys-Ct
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Alias HD-RAN PolyCys;HD-PolyCys
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the predicted polyCys frame of HD in the CTG direction (alpha-HD-Cys-Ct).
Molecular Weight of Antigen ∼50 kda observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 100 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects RAN proteins with PolyCys.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.