Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HD-polyLeu-Ct Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN167010MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN167010MI 100 μL
ABN167025MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN167010MI Supplier MilliporeSigma Supplier No. ABN1670100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HD-polyLeu-Ct Polyclonal antibody specifically detects HD-polyLeu-Ct in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HD-polyLeu-Ct
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Alias HD-RAN PolyLeu;HD-PolyLeu
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminal region of the predicted polyLeu frames of HD in the CTG direction (HD-Leu-Ct).
Molecular Weight of Antigen ∼21 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 100 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects RAN proteins with PolyLeu-HD.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.