Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HD-polySer-Ct Rabbit anti-Human,Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN167325MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN167325MI 25 μL
ABN167310MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN167325MI Supplier MilliporeSigma Supplier No. ABN167325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HD-polySer-Ct Polyclonal antibody specifically detects HD-polySer-Ct in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HD-polySer-Ct
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Alias HD-RAN-PolySer;HD-PolySer
Host Species Rabbit
Immunogen A synthetic peptides from the C-terminal regions of the predicted 21 polySer frames of HD in the CAG direction.
Molecular Weight of Antigen ∼27 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects RAN proteins with PolySer.
Target Species Human, Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.