Learn More
hD53/TPD52L1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 147321AP150UL

Description
Four variants resulted from alternative splicing have been predicated and this antibody is expected to recognize all the variants.
This gene encodes a member of the tumor protein D52 family. The encoded protein contains a coiled-coil domain and may form homo- or hetero-dimer with TPD52 family members. The protein is reported to be involved in cell proliferation and calcium signaling. It also interacts with the mitogen-activated protein kinase kinase kinase 5 and positively regulates MAP3K5-induced apoptosis. Multiple alternatively spliced transcript variants have been observed, but the full-length nature of some variants has not yet been determined.Specifications
hD53/TPD52L1 | |
Polyclonal | |
Unconjugated | |
TPD52L1 | |
D53, hD53, TDP53, TPD52L1, TPD52L2, TPD53, tumor protein D52 like 1, Tumor protein D53 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7164 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16890 | |
TPD52L1 | |
hD53; TPD52L1 Fusion Protein Ag6493 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.