Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HDAC-Glo 2 Assay, 10ml

Catalog No. PRG9590 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9590 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG9590 Supplier Promega Supplier No. G9590

The HDAC-Glo Class IIa and HDAC-Glo 2 Assays are single-reagent-addition homogeneous, luminescent assays that measure the relative activity of histone deacetylase (HDAC) Class IIa and Class I enzyme 2 from cells, extracts or purified enzyme sources.

  • Detects HDAC Class IIa or Class I Enzyme 2 Activity
  • Single−reagent−addition homogeneous, luminescent assays
  • Measures the relative activity of histone deacetylase (HDAC) Class IIa and Class I enzyme 2
  • Use cells, extracts or purified enzymes

Specifications

Quantity 10 mL
Includes 10ml
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.