Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC1 Antibody (PCRP-HDAC1-1B7), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314062AF350
This item is not returnable.
View return policy
Description
HDAC1 Monoclonal antibody specifically detects HDAC1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| HDAC1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3 | |
| Recombinant full-length human HDAC1 protein (Uniprot: Q13547) | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Loading Controls, Stem Cell Markers | |
| 3065 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-HDAC1-1B7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction