Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC1 Monoclonal Antibody (1061CT1.3.1)
HDAC1 Monoclonal Antibody (1061CT1.3.1)
Supplier: Thermo Scientific MA537620
Description
HDAC1 Monoclonal Antibody for Western Blot

Specifications
HDAC1 | |
Monoclonal | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q13547 | |
HDAC1 | |
purified His-tagged HDAC1 protein. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Western Blot | |
1061CT1.3.1 | |
Unconjugated | |
HDAC1 | |
DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 | |
Mouse | |
Euglobulin Precipitation | |
RUO | |
3065 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction