Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC1 Monoclonal Antibody (10E2), Active Motif™
SDP

Mouse Monoclonal Antibody

Supplier:  Active Motif 39532

Encompass

 View more versions of this product

Catalog No. 50-199-4035


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Histone deacetylase (HDAC) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones - H2A, H2B, H3, and H4. Histone deacetylation gives a tag for epigenetic repression and plays and important role in transcriptional regulation, cell cycle progression, and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.
Specifications

Specifications

HDAC1
Monoclonal
1 mg/mL
antibody with 30% glycerol and 0.035% sodium azide; pH
O09106, Q13547, Q4QQW4
HDAC1
This HDAC1 antibody was raised against a kLH-conjugated peptide corresponding to amino acids 467-482 of human and murine HDAC1.
RUO
297893, 3065, 433759
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
10E2
Unconjugated
HDAC1
DKFZp686H12203; GON-10; HD1; HDAC1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1
Mouse
10 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.