Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDAC1 Monoclonal Antibody (E31), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA551103
Description
Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. HDAC1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis.
Specifications
HDAC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Q13547 | |
HDAC1 | |
Recombinant Human HDAC1 protein, Met1-Ala482 (Accession #Q13547). | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
E31 | |
Unconjugated | |
HDAC1 | |
DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 | |
Mouse | |
Protein A/G | |
RUO | |
3065 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction