Learn More
HDAC1 Monoclonal Antibody (OTI5F9), TrueMAB™, OriGene
SDP
Mouse Monoclonal Antibody
Supplier:  OriGene TA502193
Encompass_Preferred
 View more versions of this product
Catalog No. 50-167-3271
Only null left
Add to Cart
Description
Description
Histone deacetylase (HDAC) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones - H2A, H2B, H3, and H4. Histone deacetylation gives a tag for epigenetic repression and plays and important role in transcriptional regulation, cell cycle progression, and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.
Specifications
Specifications
HDAC1
Monoclonal
Unconjugated
HDAC1
GON-10, HD1, RPD3, RPD3L1
Mouse
Affinity Chromatography
RUO
297893, 3065, 433759, 487309
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI5F9
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O09106, Q13547, Q4QQW4
HDAC1
Full length human recombit protein of human HDAC1 produced in HEK293T cell.
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.