Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDAC1 Polyclonal Antibody, Alexa Fluor™ 647

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1860A647

Catalog No. PIPA1860AY


Only null left
Edge
Add to Cart

Description

Description

PA1860A647 detects histone deacetylase 1 (HDAC1) from human, mouse, rat, hamster and canine tissues and cells. The PA1860A647 immunogen is a synthetic peptide corresponding to residues C E(467) E K P E A K G V K E E V K L A(482) of human HDAC1. This immunizing peptide (Cat. # PEP-069) is available for use in neutralization and control experiments.

Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. HDAC1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDAC1
Polyclonal
Alexa Fluor 647
HDAC1
DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1
Rabbit
Antigen Affinity Chromatography
RUO
100752721, 297893, 3065, 433759, 487309
4°C, store in dark, DO NOT FREEZE!
Liquid
Immunocytochemistry
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
O09106, Q13547, Q4QQW4
HDAC1
Synthetic peptide corresponding to residues C E(467) E K P E A K G V K E E V K L A(482) of human HDAC1.
50 μL
Primary
Human, Mouse, Rat, Canine, Hamster
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.