Learn More
HDAC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HDAC1 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6102
![]() ![]() |
Proteintech Group Inc
101971AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human HDAC1. The calculated molecular weight of HDAC1 is 55 kDa, but modified HDAC1 is about 60-65 kDa.
Histone deacetylase (HDAC) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones - H2A, H2B, H3, and H4. Histone deacetylation gives a tag for epigenetic repression and plays and important role in transcriptional regulation, cell cycle progression, and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.Specifications
HDAC1 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O09106, Q13547, Q4QQW4 | |
297893, 3065, 433759 | |
HDAC1 Fusion Protein Ag0256 | |
Primary | |
-20°C | |
HDAC1 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GON 10, HD1, HDAC1, histone deacetylase 1, RPD3, RPD3L1 | |
Hdac1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.