Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC1 Rabbit anti-Human, Unconjugated, Clone: AIB-8, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20120
Description
HDAC1 Monoclonal antibody specifically detects HDAC1 in Human samples. It is validated for ICC, WB-Ce, IHC-P, IF.Specifications
HDAC1 | |
Monoclonal | |
Unconjugated | |
HDAC1 | |
Rabbit | |
100 μg | |
3065 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
AIB-8 | |
histone deacetylase 1 | |
DKFZp686H12203, GON-10, HD1, RPD3, RPD3L1 | |
A synthetic peptide corresponding to human HDAC1 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction