Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC10 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600223PCP
Description
HDAC10 Polyclonal antibody specifically detects HDAC10 in Human samples. It is validated for Western Blot, Western BlotSpecifications
HDAC10 | |
Polyclonal | |
Western Blot, Simple Western | |
DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722 | |
Synthetic peptide corresponding to a portion of human HDAC 10. | |
0.1 mL | |
Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, Epigenetics, Transcription Factors and Regulators | |
83933 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Simple Western | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction