Learn More
Invitrogen™ HDAC10 Monoclonal Antibody (OTI7C3)

Description
HDAC10 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | HDAC10 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI7C3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HDAC10 |
| Gene Accession No. | Q969S8 |
| Gene Alias | AW548891; DKFZp761B039; Hd10; Hdac10; Histone deacetylase 10; MGC149722; Polyamine deacetylase HDAC10; RP3-402G11.7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.