Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDAC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550985 Supplier Invitrogen™ Supplier No. PA550985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC10 Polyclonal Antibody detects endogenous levels of HDAC10 protein.

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC10
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HDAC10
Gene Accession No. Q569C4, Q6P3E7, Q969S8
Gene Alias AW548891; DKFZp761B039; Hd10; Hdac10; Histone deacetylase 10; MGC149722; Polyamine deacetylase HDAC10; RP3-402G11.7
Gene Symbols HDAC10
Host Species Rabbit
Immunogen Synthesized peptide derived from the N-terminal region of human Histone deacetylase 10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170787, 362981, 83933
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.