Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDAC10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599170
Description
Antibody detects endogenous levels of total HDAC10.
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.
Specifications
HDAC10 | |
Polyclonal | |
Unconjugated | |
HDAC10 | |
AW548891; DKFZp761B039; Hd10; Hdac10; Histone deacetylase 10; MGC149722; Polyamine deacetylase HDAC10; RP3-402G11.7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
170787, 362981, 83933 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q569C4, Q6P3E7, Q969S8 | |
HDAC10 | |
A synthesized peptide derived from human HDAC10(Accession Q969S8), corresponding to amino acid residues A4-E54. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction