Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDAC10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144723
Description
Immunogen sequence: LTTPDITLVL PPDVIQQEAS ALREETEAWA RPHESLAREE ALTALGKLLY LLDGMLDGQV NSGIAATPAS AAAATLDVAV RRGLSHGAQR LLCVALGQLD RPPDLAHDGR SLWLNIRGKE AAALSMFHVS Antigen sequence identity: Mouse 64%; Rat 51%.
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.
Specifications
| HDAC10 | |
| Polyclonal | |
| Unconjugated | |
| HDAC10 | |
| AW548891; DKFZp761B039; Hd10; Hdac10; Histone deacetylase 10; MGC149722; Polyamine deacetylase HDAC10; RP3-402G11.7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 83933 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969S8 | |
| HDAC10 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 416-545. Recombinant protein control fragment (Product #RP-110180). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction