Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC11 Antibody, Novus Biologicals™
SDP

Catalog No. p-5071157 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056382T 0.025 mg
NB10056382 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056382T Supplier Novus Biologicals Supplier No. NB10056382SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC11 Polyclonal specifically detects HDAC11 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen HDAC11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Simple Western reported by customer review
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9UKV0
Gene Alias EC 3.5.1.98, FLJ22237, HD11, histone deacetylase 11
Gene Symbols HDAC11
Host Species Rabbit
Immunogen This antibody was developed by immunizing mice with a synthetic peptide (AHQGNGHERDFMDDKRVYI) of human HDAC11.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 79885
Test Specificity This peptide sequence is 100% homologous in multiple HDAC11 isoforms corresponding to amino acids 60-78. The observed molecular weight in western blots can vary depending on the isoform(s) detected. Users are encouraged to refer to NBCI data bases for more information about HDAC-11 isoforms and their amino acid size.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.