Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC11 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298764PCP
Description
HDAC11 Polyclonal antibody specifically detects HDAC11 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
HDAC11 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EC 3.5.1.98, FLJ22237, HD11, histone deacetylase 11 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human HDAC11. | |
0.1 mL | |
Breast Cancer, Cancer, Epigenetics | |
79885 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction