Learn More
Invitrogen™ HDAC11 Polyclonal Antibody

Description
This peptide sequence is 100% homologous in multiple HDaC11 isoforms. Suggested positive control: human, mouse, or rat brain. Immunogen sequence: AHQGNGHERD FMDDKRVYI.
Specifications
Specifications
| Antigen | HDAC11 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | HDAC11 |
| Gene Accession No. | Q91WA3, Q96DB2 |
| Gene Alias | FLJ22237; HD11; HDAC11; Histone deacetylase 11 |
| Gene Symbols | HDAC11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.