Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC11 Rabbit anti-Human, Mouse, Rat, Unconjugated, Polyclonal, Active Motif™
SDP

Catalog No. 501992652
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2652 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2652 Supplier Active Motif Supplier No. 39208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a class IV histone deacetylase. The encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10. Alternative splicing results in multiple transcript variants.

Specifications

Antigen HDAC11
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide; pH 7.5
Gene HDAC11
Gene Accession No. Q91WA3, Q96DB2
Gene Alias FLJ22237; HD11; Hdac11; Histone deacetylase 11
Gene Symbols HDAC11
Host Species Rabbit
Immunogen This HDAC11 antibody was raised against a peptide corresponding to amino acid residues 238-251 of human HDAC11.
Purification Method Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232232, 297453, 79885
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.