Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC2 Monoclonal Antibody (3F3), Active Motif™

Mouse Monoclonal Antibody
$156.65 - $638.65
Specifications
Antigen | HDAC2 |
---|---|
Clone | 3F3 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-4037
![]() ![]() |
Active Motif
39534 |
10 μL |
Each for $156.65
|
|
|||||
50-199-4036
![]() ![]() |
Active Motif
39533 |
200 μL |
Each for $638.65
|
|
|||||
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein shows sequence similarity to the E. coli RNA polymerase-binding protein HepA. Mutations in this gene are a cause of Schimke immunoosseous dysplasia (SIOD), an autosomal recessive disorder with the diagnostic features of spondyloepiphyseal dysplasia, renal dysfunction, and T-cell immunodeficiency.Specifications
HDAC2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
antibody with 30% glycerol and 0.035% sodium azide; pH | |
BOS_9605; D10Wsu179e; HD2; HDAC2; Histone deacetylase 2; histone deacetylase 2-like protein; histone deacetylase-2; I79_010219; mRPD3; RPD3; transcriptional regulator homolog RPD3; YAF1; YY1 transcription factor-binding protein; YY1-associated factor 1; Yy1bp | |
HDAC2 | |
IgG1 | |
-20°C, Avoid Freeze/Thaw Cycles | |
HDAC2 |
3F3 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P70288, Q92769 | |
15182, 3066, 84577 | |
This HDAC2 antibody was raised against a kLH-conjugated peptide corresponding to amino acids 473-488 of human HDAC2. | |
Primary | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title