Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC2 Monoclonal Antibody (7G5), Bioss
SDP

Catalog No. 502279818
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9818 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9818 Supplier Bioss Supplier No. BSM53026M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HDAC2 Monoclonal antibody specifically detects HDAC2 in Human samples. It is validated for ChIP assay, ChIP-sequencing, Dot Blot, ELISA, Immunocytochemistry, Western Blot

Specifications

Antigen HDAC2
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7G5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene HDAC2
Gene Accession No. Q92769
Gene Alias D10Wsu179e; HD2; hdac 2; Hdac2; hdac-2; Histone deacetylase 2; histone deacetylase 2-like protein; histone deacetylase-2; I79_010219; mRPD3; RPD3; transcriptional regulator homolog RPD3; YAF1; YY1 transcription factor-binding protein; YY1-associated factor 1; Yy1bp
Host Species Mouse
Immunogen Monoclonal antibody raised in mouse against human HDAC2 (Histone deacetylase 2), using a KLH-conjugated synthetic peptide containing a sequence from the C-terminal region of the protein
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3066
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.