Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC2 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Mouse Recombinant Monoclonal Antibody
$108.45 - $482.00
Specifications
Antigen | HDAC2 |
---|---|
Concentration | 1 mg/mL |
Applications | ChIP sequencing (ChIP-seq), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3733
![]() ![]() |
Active Motif
91198 |
10 μg |
Each for $108.45
|
|
|||||
50-199-3736
![]() ![]() |
Active Motif
91197 |
100 μg |
Each for $482.00
|
|
|||||
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein shows sequence similarity to the E. coli RNA polymerase-binding protein HepA. Mutations in this gene are a cause of Schimke immunoosseous dysplasia (SIOD), an autosomal recessive disorder with the diagnostic features of spondyloepiphyseal dysplasia, renal dysfunction, and T-cell immunodeficiency.Specifications
HDAC2 | |
ChIP sequencing (ChIP-seq), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q92769 | |
3066 | |
This antibody was raised against peptide corresponding to amino acids 473-488 of human HDAC2. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
HDAC2 |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 35mM sodium acetate with 30% glycerol, 70mM NaCl and 0.07% sodium azide; pH 7.5 | |
BOS_9605; D10Wsu179e; HD2; HDAC2; Histone deacetylase 2; histone deacetylase 2-like protein; histone deacetylase-2; I79_010219; mRPD3; RPD3; transcriptional regulator homolog RPD3; YAF1; YY1 transcription factor-binding protein; YY1-associated factor 1; Yy1bp | |
HDAC2 | |
IgG2a | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title