Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$206.50 - $501.50
Specifications
Antigen | HDAC3 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 - 5ug/ml, Simple Western 1:25, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1001669SS
![]() |
Novus Biologicals
NB1001669SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB1001669
![]() |
Novus Biologicals
NB1001669 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
HDAC3 Polyclonal specifically detects HDAC3 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
HDAC3 | |
Western Blot 2 - 5ug/ml, Simple Western 1:25, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 | |
HDAC3 | |
IgG | |
Protein G purified | |
In HeLa, a 50 kDa band is observed. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases | |
PBS with 0.05% Sodium Azide | |
8841 | |
This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 2-17 of human HDAC3. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title