Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC3 Antibody (PCRP-HDAC3-2D4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314060
This item is not returnable.
View return policy
Description
HDAC3 Monoclonal antibody specifically detects HDAC3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
HDAC3 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1 to 2 μg /million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunoprecipitation 1 to 2 μg per 100-500 μg protein, Protein Array | |
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 | |
Recombinant full-length human HDAC3 protein (Uniprot: O15379) | |
100 μg | |
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics | |
8841 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG2b |
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
PCRP-HDAC3-2D4 | |
Unconjugated | |
10mM PBS without Preservative | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction