Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC3 Antibody (PCRP-HDAC3-3C9), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314061MFV610
Description
HDAC3 Monoclonal antibody specifically detects HDAC3 in Human samples. It is validated for Flow CytometrySpecifications
HDAC3 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-HDAC3-3C9 | |
Flow Cytometry | |
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 | |
Recombinant full-length human HDAC3 protein (Uniprot: O15379) | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics | |
8841 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction